missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Hyaluronidase (Bovine)

Product Code. 16181770
Click to view available options
Quantity:
50,000 U
Unit Size:
50000 units
This item is not returnable. View return policy

Product Code. 16181770

Brand: Abnova™ P5233.50000Units

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 55kDa
Name Hyaluronidase (Bovine)
Preparation Method Native protein purified from Bovine Testes
Quantity 50,000 U
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name HYALURONIDASE
Assays Hyaluronidase Assay
Biological Activity ≥300 USP/NF units per mg dry weight. One Unit equals a U.S.P./National Formulary Unit and is referenced to a standard NF Hyaluronidase.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Testes
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.