Learn More
Boekel Scientific™ Hybridization Glass Bottles for Boekel Little Shot Oven
Product Code. 10475743
Change view
Click to view available options
Length (Metric):
100 mm
150 mm
225 mm
300 mm
Unit Size:
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10475743 225 mm Pack of 2
10465743 100 mm Pack of 2
10784994 150 mm Pack of 2
10345393 300 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10475743 Supplier Boekel Scientific™ Supplier No. 5020225
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Boekel Little Shot Oven, these glass hybridization bottles fit the standard carousel and help maintain the perfect temperature and mixing for all applications.
  • Pack of 2 Bottles
  • Comes with Blue Caps
  • For Use with Standard Carousel
Specifications
Color Blue
Diameter (English) Inner 1.37 in.
Diameter (Metric) Inner 35 mm
For Use With (Equipment) Boekel Little SHOT III Medium Capacity Oven
Includes Blue caps and silicone O-ring
Length (English) 8.85 in.
Length (Metric) 225 mm
Material Glass
Product Type Hybridization Bottle