Learn More
Cytiva HyClone™ Human Mesenchymal Stem Cell Screened Characterized Fetal Bovine Serum, US Origin
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
Human Mesenchymal Stem Cell Screened FBS supports the growth of undifferentiated human mesenchymal stem cells (hMSC). During screening, hMSC are observed for evidence of nutritional deficiency, cytotoxicity, or morphological aberrations.
Specifications
Specifications
| Country of Origin | United States |
| Form | Liquid |
| Specialty Type | Human mesenchymal stem cell screened |
| Quantity | 500 mL |
| Sterilization Method | 40 nm filtration |
| Endotoxin Concentration | ≤ 10 EU/mL |
| Heat Inactivated | No |
| Hemoglobin Concentration | ≤ 10 mg/dL |