Learn More
(-)-Hycosamine, TRC
Product Code. 30138502
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138502 1 g 1g
30132881 5 g 5g
18092310 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138502 Supplier Toronto Research Chemicals Supplier No. TRCH6743001G
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101-31-5
Molecular Formula C17 H23 N O3
Molecular Weight (g/mol) 289.37
Synonym 1αH,5αH-Tropan-3α-ol, (-)-tropate (ester) (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, [3(S)-endo]-, Tropic acid, 1αH,5αH-tropan-3α-yl ester, (-)- (8CI), (-)-Atropine, (-)-Hyoscyamine, (S)-(-)-Hyoscyamine, (S)-Atropine, 1αH,5αH-Tropan-3α-yl (-)-tropate, Cystospaz, Daturine, Duboisine, Hyoscyamine, L-Hyoscyamin, L-Hyoscyamine, l-Atropine, l-Hyoscyamine, l-Tropine tropate, Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, (αS)-
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2S)-3-hydroxy-2-phenylpropanoate
SMILES CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](CO)c3ccccc3
Specifications
Quantity 1 g
InChI Formula InChI=1S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19H,7-11H2,1H3/t13-,14+,15+,16-/m1/s1
Formula Weight 289.1678
Percent Purity >95%
Chemical Name or Material Hyoscyamine
Grade HPLC