missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HYDIN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566899
This item is not returnable.
View return policy
Description
Immunogen sequence: YLPGVPEVFKR SFQIQIAHLD PENITLSGEG IFPQICLDLP RNLTANEKYE MFLNQARKNT DKEYNKCEML DHFDVITEEV PEDEPAEVSA HLQ Highest antigen sequence identity to the following orthologs - mouse 69%, rat 71%.
Required for ciliary motility.
Specifications
| HYDIN | |
| Polyclonal | |
| Unconjugated | |
| HYDIN | |
| CILD5; HYDIN; HYDIN, axonemal central pair apparatus protein; HYDIN1; HYDIN2; Hydrocephalus-inducing protein homolog; KIAA1864; PPP1R31; protein phosphatase 1, regulatory subunit 31 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54768 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4G0P3 | |
| HYDIN | |
| Recombinant Human HYDIN. Recombinant protein control fragment (Product #RP-107549). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction