missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Standard: N2H4 @ 100 mg/L in 1% Acetic Acid, VHG™

Product Code. 30003169
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 30003169

Brand: VHG Labs VHGIHYD100500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Hydrazine Standard: N2H4 @ 100 mg/L in 1% Acetic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrazine
CAS 302-01-2
Methods Wet Chemistry
Analyte or Component Names Hydrazine
Shipping Condition Room Temperature
Molecular Formula N2H4
Synonym N2H4
SMILES NN
IUPAC Name Hydrazine
Solvent or Matrix Dilute Acetic Acid
Molecular Weight (g/mol) 32.05
Quantity 500 mL
Formula Weight 32.05 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.