missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine Standard: N2H4 @ 1000 mg/L in 1% Acetic Acid, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Hydrazine Standard: N2H4 @ 1000 mg/L in 1% Acetic Acid
Specifications
Specifications
| Chemical Name or Material | Hydrazine |
| CAS | 302-01-2 |
| Methods | Wet Chemistry |
| Analyte or Component Names | Hydrazine |
| Shipping Condition | Room Temperature |
| Molecular Formula | N2H4 |
| Synonym | N2H4 |
| SMILES | NN |
| IUPAC Name | Hydrazine |
| Solvent or Matrix | Dilute Acetic Acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction