missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear Hydro-Tec™ Stability Inlay Soles
Provides foot with side support and extra support at heel and middle of foot.
Brand: Emma Safety Footwear 1072/44
Additional Details : Weight : 0.01000kg
Description
- Regulates moisture
- Contains active carbon to prevent odor
Specifications
Gray | |
44 | |
Stability Inlay Sole |