missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 6.0N Standardized Solution, Thermo Scientific Chemicals

Catalog Number p-4781933
30.80 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 L
4 L
20 L
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7647-01-0, 7732-18-5
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
chlorane
Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7647-01-0, 7732-18-5
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
chlorane
Cl

6.0N HClsolution acts as a standard solution in volumetric titrations. In addition, it has industrial applications such as chemical transformations in hydrochlorination of rubber and production of vinyl and alkyl halide. In the semiconductor industry, it is used to etch semiconductor crystal and to purify silicon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 L
Color Colorless to Yellow
Formula Weight 36.46
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 49.6%; Water: 50.4%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Miscible with water, methanol,ethanol and ether
Physical Form Liquid
Chemical Name or Material Hydrochloric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'