Learn More
Hydrochloric Acid Solution, 0.2M, Chem Lab™
Catalog Number p-8014299
Click to view available options
Quantity:
5 L
Packaging:
2Each
946mL
Appearance Liquid
pH <1
Appearance of Solution Liquid
Color Colorless
Composition 7.292g HCl/l H2O
Concentration 7.292g HCl/l H2O = 0.2 N (±0.0004/20°C)
Density 1.01g/mL
Odor Odourless
Packaging 2Each
Quantity 5 L
Physical Form Liquid
Solubility Information Soluble in water
Show More Show Less
compliance-icons
Product Identifier
  • Hydrochloric acid 0.2 mol/l
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid