missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, nominally 2.5M in ethanol, Thermo Scientific Chemicals

Catalog Number p-7052200
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
Cl
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
Cl

Hydrogen chloride, 2.5M. in ethanol is used in the manufacturing of pharmaceuticals to get a hydrochloride salt. For example, ranitidine base is converted to ranitidine hydrochloride, which is used as anti ulcerative drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN2924
Concentration Nominally 2.5M in ethanol
Solubility Information Miscible with water at room temperature. Also miscible with methanol,ethanol and ether.
IUPAC Name chlorane
Formula Weight 36.46
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Hydrochloric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'