missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Hydrogen Peroxide Assay Kit (Fluorometric)
Shop All Bio Techne Products
Click to view available options
Quantity:
96 Tests
Unit Size:
1 set
Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Quantity | 96 Tests |
| Species | Non-species specific |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction