missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide Solution, 30%, Chem-Lab
Description
Hydrogen peroxide’s oxidizing properties are so strong that it is considered a highly reactive oxygen species. Due to it being a weak acid, it can form hydroperoxide or peroxide salts. Industrially, it is used for pulp/paper bleaching, in the manufacture of sodium percarbonate and sodium perborate, and in water purification to remove organic impurities and odors.
Specifications
Specifications
| Color | Colorless |
| CAS | 7722-84-1 |
| Boiling Point | 107°C |
| Melting Point | -26°C |
| Molecular Formula | H2O2 |
| Composition | 30+% H2O2 |
| Density | 1.11g/mL |
| Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
| Solubility Information | Water Soluble |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| Show More |
Safety and Handling
Product Identifier
- Hydrogen peroxide 30 weight % solution a.r.
- Danger
- Oxidising liquid Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contain: Hydrogen peroxide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?