missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Potassium ATPase Beta Antibody (SR1980), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22395-50ul
This item is not returnable.
View return policy
Description
Hydrogen Potassium ATPase Beta Monoclonal antibody specifically detects Hydrogen Potassium ATPase Beta in Human samples. It is validated for Western Blot
Specifications
| Hydrogen Potassium ATPase Beta | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1980 | |
| Western Blot 1:500-1:2000 | |
| ATP6B, ATPase, H+/K+ exchanging, beta polypeptide, ATPase, H+/K+ transporting, beta polypeptide, Gastric H(+)/K(+) ATPase subunit beta, gastric H+/K+ ATPase beta subunit, gastric hydrogen-potassium ATPase, beta, potassium-transporting ATPase beta chain, potassium-transporting ATPase subunit beta, Proton pump beta chain | |
| A synthesized peptide derived from human Hydrogen Potassium ATPase Beta (Uniprot #: P51164) | |
| 50 μg | |
| Endocrinology, Signal Transduction | |
| 496 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction