Learn More
Brannan™ Low Tension Specific Gravity Hydrometer
Click to view available options
Length (Metric):
270 mm
Unit Size:
Each
Description
The specific gravity 60/60°F scale compares the relative density of a given liquid with that of water, at 60°F (15.6°C).
- Manufactured from soda glass, steel shot ballast and synthetic wax resin
- Range of 0.050 S60/60°F
- M50, 270mm long
- Low tension 0.90 to 0.95
Specifications
Specifications
| Increments | 0.001 S60/60°F |
| Length (Metric) | 270 mm |
| Temperature Accuracy | 0.001 S60/60°F |
| Material | Soda glass, steel shot ballast, synthetic wax resin |
| Temperature Range (English) | 0.050 S60/60°F |
| Ballast Type | Steel Shot |
| For Use With (Application) | Comparing relative density of a liquid to water |