missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hydromorphone Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA175167
This item is not returnable.
View return policy
Description
Working Concentration: 1.25 μg/mL. Sensitivity: 10 ng/mL Hydromorphone produces 74% inhibition in a competitive ELISA employing Hydromorphone polyclonal antibody.
Hydromorphone Base is the hydrogenated ketone of morphine, a semi-synthetic opioid with analgesic effects. Hydromorphone selectively binds the mu-opioid receptor which is linked through G-proteins. Binding stimulates the exchange of guanosine triphosphate (GTP) for guanosine diphosphate (GDP) on the G-protein complex and interacts with and inhibits adenylate cyclase located at the inner surface of the plasma membrane. This leads to a reduction in intracellular cyclic 3',5'-adenosine monophosphate (cAMP). Further, voltage-gated potassium channels are activated, thereby causing hyperpolarization and reducing neuronal excitability. In addition, the opening of voltage-gated calcium channels is inhibited, thereby leading to an inhibition of calcium entry and a reduction in the release of various neurotransmitters, including GABA, vasopressin, somatostatin, insulin and glucagons.
Specifications
| Hydromorphone | |
| Polyclonal | |
| Unconjugated | |
| C17H19NO3; Dihydromorfinon; Dihydromorphinone; Dilaudid; Dimorphone; Idromorfone | |
| Hydromorphone(3)-BTG. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA | |
| 7.28 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.8 | |
| Sheep | |
| purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction