Learn More
Corning™ Stripettor™ Ultra Pipet Controller Accessory, Hydrophobic PTFE Filters
Extreme chemical resistance of PTFE membranes makes them very useful for filtering solvents or other aggressive chemicals for which other membranes are unsuitable.
Brand: Corning™ 4992
Additional Details : Weight : 0.20000kg
Alert:
Because of their hydrophobicity, PTFE membranes must be prewetted with a solvent, such as ethanol, before aqueous solutions can be filtered.
Description
These filters are use in the Corning™ StripettorTM Ultra Pipet Controller.
Specifications
White | |
Hydrophobic PTFE Filter | |
Stripettor™ Ultra Pipet Controller |
0.45 μm | |
Polytetrafluorethylene (PTFE) | |
5/Pk. |