missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxy Tyrosol (>85%), TRC
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
Specifications
Specifications
| Chemical Name or Material | Hydroxytyrosol |
| CAS | 10597-60-1 |
| Molecular Formula | C8 H10 O3 |
| Quantity | 1 g |
| Synonym | Phenethyl alcohol, 3,4-dihydroxy- (6CI,7CI,8CI), 4-(2-Hydroxyethyl)-1,2-benzenediol, 1-(2-Hydroxyethyl)-3,4-dihydroxybenzene, 2-(3,4-Dihydroxyphenyl)ethanol, 2-(3,4-Dihydroxyphenyl)ethyl alcohol, 3,4-DHPEA, 3,4-Dihydroxy-β-phenethyl alcohol, 3,4-Dihydroxybenzeneethanol, 3,4-Dihydroxyphenethyl alcohol, 3,4-Dihydroxyphenylethanol, 3,4-Dihydroxyphenylethyl alcohol, 3-Hydroxytyrosol, Ba 2774, Homoprotocatechuyl alcohol, Hydroxytyrosol, β-(3,4-Dihydroxyphenyl)ethanol, β-(3,4-Dihydroxyphenyl)ethyl alcohol |
| InChI Formula | InChI=1S/C8H10O3/c9-4-3-6-1-2-7(10)8(11)5-6/h1-2,5,9-11H,3-4H2 |
| SMILES | OCCc1ccc(O)c(O)c1 |
| IUPAC Name | 4-(2-hydroxyethyl)benzene-1,2-diol |
| Molecular Weight (g/mol) | 154.16 |
| Formula Weight | 154.063 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction