missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyacid Oxidase-1/HAO-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99928-100ul
This item is not returnable.
View return policy
Description
Hydroxyacid Oxidase-1/HAO-1 Polyclonal antibody specifically detects Hydroxyacid Oxidase-1/HAO-1 in Human samples. It is validated for ELISA
Specifications
| Hydroxyacid Oxidase-1/HAO-1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| (S)-2-hydroxy-acid oxidase, EC 1.1.3.15, Glycolate oxidase, GOX1MGC142227, GOXMGC142225, HAOX1, hydroxyacid oxidase (glycolate oxidase) 1, hydroxyacid oxidase 1 | |
| Produced in rabbits immunized with purified, recombinant Human Hydroxyacid Oxidase-1/HAO-1 (Accession#: NP_060015.1; Leu 2-Ile 370) | |
| 100 μg | |
| metabolism, Signal Transduction | |
| 54363 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction