missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyacid Oxidase-1/HAO-1 Antibody (OTI5C3), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71954C
This item is not returnable.
View return policy
Description
Hydroxyacid Oxidase-1/HAO-1 Monoclonal specifically detects Hydroxyacid Oxidase-1/HAO-1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Hydroxyacid Oxidase-1/HAO-1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HAO1 | |
| Full length recombinant protein of human Hydroxyacid Oxidase-1/HAO-1 (NP_060015) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| OTI5C3 | |
| Western Blot, Flow Cytometry | |
| (S)-2-hydroxy-acid oxidase, EC 1.1.3.15, Glycolate oxidase, GOX1MGC142227, GOXMGC142225, HAOX1, hydroxyacid oxidase (glycolate oxidase) 1, hydroxyacid oxidase 1 | |
| Mouse | |
| Affinity Purified | |
| metabolism, Signal Transduction | |
| 54363.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction