missing translation for 'onlineSavingsMsg'
Learn More

Hydroxysafflor yellow A, MedChemExpress

Product Code. 16479018
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479018 10 mg 10mg
16424857 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479018 Supplier Medchem Express Supplier No. HYN0567

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hydroxysafflor yellow A is a flavonoid derived and isolated from traditional Chinese medicine Carthamus tinctorius L.

Hydroxysafflor yellow A is a flavonoid derived and isolated from traditional Chinese medicine Carthamus tinctorius L.

Specifications

Chemical Name or Material Hydroxysafflor yellow A
CAS 78281-02-4
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C27H32O16
Quantity 10 mg
Synonym Safflomin A HSYA
Solubility Information DMSO : 250 mg/mL (408.14 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (54.41 mM; Need ultrasonic)
SMILES OC(C(O)=C1C(/C=C/C2=CC=C(O)C=C2)=O)([C@]([C@@H]([C@@H](O)[C@@H]3O)O)([H])O[C@@H]3CO)C(O)=C([C@@H]([C@@H]([C@@H](O)[C@@H]4O)O)O[C@@H]4CO)C1=O
Molecular Weight (g/mol) 612.53
Formula Weight 612.53
Percent Purity 95.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.