Learn More
Ansell™ HyFlex™ 11-500 Series Nitrile Medium Weight Gloves
Designed for dry or slightly oily applications where workers need light cut protection or isolation from heat or flying molten metal debris.
Brand: Ansell™ 11500110
Additional Details : Weight : 0.63900kg
Description
- Knitted construction
- Nylon Lycra™ Kevlar™ liner
- Nitrile coating
- Palm-coated finish
- Knitwrist cuff
- Black and yellow color