Learn More
Invitrogen™ HYI Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588767
Description
Immunogen sequence: PGRQAAFREG LEQAVRYAKA LGCPRIHLMA GRVPQGADRI AVKAEMEAVF LENLRHAAGV LAQEDLVGLL EPINTRITDP QYFLDTPQQA AAILQKVGRP NLQLQMDIFH WQIMDGNLTG NIREFLPIVG HVQVAQVPGR GEPSSPGELN FPYLFQLLED EGYKGFVGCE YQPRGDTVEG L; Positive Samples: Mouse liver, Mouse lung, Rat spleen.
Catalyzes the reversible isomerization between hydroxypyruvate and 2-hydroxy-3-oxopropanoate (also termed tartronate semialdehyde).
Specifications
| HYI | |
| Polyclonal | |
| Unconjugated | |
| Hyi | |
| 2700033B16Rik; 6430559E15Rik; endothelial cell apoptosis protein E-CE1; HT036; hydroxypyruvate isomerase; hydroxypyruvate isomerase (putative); hydroxypyruvate isomerase homolog; hydroxypyruvate isomerase homolog (E. coli); HYI; Putative hydroxypyruvate isomerase; RGD1561416; SB156 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 500536, 68180, 81888 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5T013, Q8R1F5 | |
| Hyi | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-260 of human HYI (NP_0011778091). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.