missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HYLS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117996
This item is not returnable.
View return policy
Description
HYLS1 Polyclonal Antibody for Western Blot, IHC (P), IP
This gene encodes a protein localized to the cytoplasm. Mutations in this gene are associated with hydrolethalus syndrome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| HYLS1 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q96M11 | |
| HYLS1 | |
| E. coli-derived Human HYLS1 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| HYLS1 | |
| 3010015K02Rik; HLS; hydrolethalus syndrome 1; hydrolethalus syndrome protein 1; Hydrolethalus syndrome protein 1 homolog; Hyls1; HYLS1 centriolar and ciliogenesis associated; HYLS1, centriolar and ciliogenesis associated; hypothetical LOC538735 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 219844 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction