missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYPB Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-77334
This item is not returnable.
View return policy
Description
HYPB Polyclonal specifically detects HYPB in Human samples. It is validated for Flow Cytometry, Peptide ELISA.
Specifications
| HYPB | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FLJ22472, FLJ23184, FLJ45883, FLJ46217, HIF1, HIF-1EC 2.1.1.43, HIP-1, histone-lysine N-methyltransferase SETD2, hSET2, huntingtin interacting protein 1, Huntingtin yeast partner B, Huntingtin-interacting protein 1, Huntingtin-interacting protein B, HYPBp231HBP, KIAA1732SET domain-containing protein 2, KMT3AHBP231, Lysine N-methyltransferase 3A, SET domain containing 2, SET2FLJ16420 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29072 | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, KnockDown | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 2 ug/mL, Flow Cytometry 10 ug/mL, Peptide ELISA Detection limit 1:128000, Knockdown Validated | |
| NP_054878.3 | |
| SETD2 | |
| Peptide with sequence C-ERDPDKQTQNKE corresponding to internal region according to NP_054878.5. | |
| 0.1 mg | |
| Primary | |
| Expected from sequence similarity: Human, Dog, Pig. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction