missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYPE Antibody (OTI2A11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45664
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
HYPE Monoclonal specifically detects HYPE in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| HYPE | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 | |
| AMPylator FICD, EC 2.7.7.n1, FIC domain containing, FIC domain-containing protein, fic S-phase protein cell division homolog, HIP-13, HIP13UNQ3041, huntingtin interacting protein 13, Huntingtin interacting protein E, huntingtin interactor protein E, Huntingtin yeast partner E, Huntingtin-interacting protein 13, Huntingtin-interacting protein E, HYPEadenosine monophosphate-protein transferase FICD, MGC5623 | |
| Mouse | |
| 51.6 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 11153 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI2A11 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| FICD | |
| Human recombinant protein fragment corresponding to amino acids 61-265 of human FICD (NP_009007) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction