Learn More
Terumo™ Agani™ Single-use Sterile Hypodermic Needles
Click to view available options
Needle Gauge:
18 G
19 G
20 G
21 G
22 G
23 G
24 G
25 G
26 G
27 G
30 G
Color:
Black
Blue
Brown
Gray
Green
Ivory
Maroon
Orange
Pink
Purple
Yellow
Needle Point Style:
11° Regular
11° Triple Bevel
17° Short
Length (Metric):
13 mm
16 mm
19 mm
23 mm
25 mm
32 mm
38 mm
50 mm
Unit Size:
Pack of 100
Description
- Constructed from polypropylene, stainless steel, epoxy resin and dimethyl silicone oil
- All needles are supplied sterile wrapped in individual, tamper proof peel pouches
- Simple color coded system allows easy identification of required sizes
- Use with a wide range of Sterile Syringes
- Needles are sterilized with ethylene oxide to a Sterility Assurance Level of at least 10-6 Class IIa, sterile product following MDD 93/42/EEC
- CE marked and latex free
- Supplied in boxes of 100 needles
Specifications
Specifications
| Material | Stainless Steel |
| Color | Maroon |
| Needle Gauge | 26 G |
| Diameter (Metric) Outer | 0.45 mm |
| Length (Metric) | 13 mm |
| Product Type | Sterile Hypodermic Needle for Single-Use |
| Needle Point Style | 11° Triple Bevel |
| Sterility | Sterile |