Learn More
- Home
- Products
- First Aid and Medical
- Patient Care Products
- Medical Syringes
- Terumo™ Agani™ Single-use Sterile Hypodermic Needles
Terumo™ Agani™ Single-use Sterile Hypodermic Needles
Product Code. 15321537
Change view
Click to view available options
Needle Gauge:
18 G
19 G
20 G
21 G
22 G
23 G
24 G
25 G
26 G
27 G
30 G
Color:
Black
Blue
Brown
Gray
Green
Ivory
Maroon
Orange
Pink
Purple
Yellow
Needle Point Style:
11° Regular
11° Triple Bevel
17° Short
Length (Metric):
13 mm
16 mm
19 mm
23 mm
25 mm
32 mm
38 mm
50 mm
Unit Size:
Pack of 100
25 product options available for selection
| Product Code. | Needle Gauge | Color | Needle Point Style | Length (Metric) | Unit Size |
|---|---|---|---|---|---|
| 15321537 | 18 G | Pink | 11° Regular | 38 mm | Pack of 100 |
| 15351557 | 18 G | Pink | 17° Short | 38 mm | Pack of 100 |
| 15331537 | 19 G | Ivory | 11° Regular | 25 mm | Pack of 100 |
| 15341537 | 19 G | Ivory | 11° Regular | 38 mm | Pack of 100 |
| 15361557 | 19 G | Ivory | 17° Short | 38 mm | Pack of 100 |
| 15371537 | 21 G | Green | 11° Regular | 16 mm | Pack of 100 |
| 15438652 | 21 G | Green | 11° Regular | 50 mm | Pack of 100 |
| 15301547 | 22 G | Black | 11° Regular | 32 mm | Pack of 100 |
| 15331547 | 23 G | Blue | 11° Regular | 16 mm | Pack of 100 |
| 15351547 | 23 G | Blue | 11° Regular | 32 mm | Pack of 100 |
| 15361547 | 24 G | Purple | 11° Regular | 25 mm | Pack of 100 |
| 15391547 | 25 G | Orange | 11° Regular | 25 mm | Pack of 100 |
| 15331557 | 27 G | Gray | 11° Regular | 16 mm | Pack of 100 |
| 15391557 | 30 G | Yellow | 11° Regular | 13 mm | Pack of 100 |
| 15311557 | 26 G | Brown | 11° Regular | 23 mm | Pack of 100 |
| 15301557 | 26 G | Maroon | 11° Triple Bevel | 13 mm | Pack of 100 |
| 15371547 | 25 G | Orange | 11° Triple Bevel | 16 mm | Pack of 100 |
| 15361537 | 20 G | Yellow | 11° Triple Bevel | 38 mm | Pack of 100 |
| 15321547 | 22 G | Black | 11° Triple Bevel | 38 mm | Pack of 100 |
| 15341547 | 23 G | Blue | 11° Triple Bevel | 25 mm | Pack of 100 |
| 15341557 | 27 G | Gray | 11° Triple Bevel | 19 mm | Pack of 100 |
| 15391537 | 21 G | Green | 11° Triple Bevel | 38 mm | Pack of 100 |
| 15381537 | 21 G | Green | 11° Regular | 25 mm | Pack of 100 |
| 15354468 | 23 G | Blue | 11° Regular | 25 mm | Pack of 100 |
| 15351537 | 20 G | Yellow | 11° Regular | 25 mm | Pack of 100 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
This item is not returnable.
View return policy
Product Code.
15321537
Supplier
Terumo™
Supplier No.
8AN1838R1
You must Sign In and have a business account to purchase this item.
Restricted Item
In Stock
Log in to see stock.
Log in to see stock.
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable.
View return policy
Description
- Constructed from polypropylene, stainless steel, epoxy resin and dimethyl silicone oil
- All needles are supplied sterile wrapped in individual, tamper proof peel pouches
- Simple color coded system allows easy identification of required sizes
- Use with a wide range of Sterile Syringes
- Needles are sterilized with ethylene oxide to a Sterility Assurance Level of at least 10-6 Class IIa, sterile product following MDD 93/42/EEC
- CE marked and latex free
- Supplied in boxes of 100 needles
Specifications
Specifications
| Color | Pink |
| Needle Gauge | 18 G |
| Diameter (Metric) Outer Needle | 1.2 mm |
| Length (Metric) | 38 mm |
| Product Type | Sterile Hypodermic Needle for Single-Use |
| Needle Point Style | 11° Regular |
| Sterility | Sterile |