missing translation for 'onlineSavingsMsg'
Learn More

I-BET762 carboxylic acid, MedChemExpress

Product Code. 16416837
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16416837

Brand: Medchem Express HY107443

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress I-BET762 carboxylic acid (Molibresib carboxylic acid) is an I-BET762-based warhead ligand for conjugation reactions of PROTAC targeting on BET. I-BET762 carboxylic acid (Molibresib carboxylic acid) is a BRD4 inhibitor with a pIC50 of 5.1.

I-BET762 carboxylic acid (Molibresib carboxylic acid) is an I-BET762-based warhead ligand for conjugation reactions of PROTAC targeting on BET. I-BET762 carboxylic acid (Molibresib carboxylic acid) is a BRD4 inhibitor with a pIC50 of 5.1.

Specifications

Chemical Name or Material I-BET762 carboxylic acid
CAS 1300019-38-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H17ClN4O3
Quantity 5 mg
Synonym Molibresib carboxylic acid GSK525762A carboxylic acid PROTAC BRD4-binding moiety 2
Solubility Information DMSO : 7.14 mg/mL (17.99 mM; Need ultrasonic)
SMILES CC1=NN=C2[C@H](CC(O)=O)N=C(C3=CC=C(Cl)C=C3)C4=C(C=CC(OC)=C4)N21
Molecular Weight (g/mol) 396.83
Formula Weight 396.83
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.