missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Burkholderia cepacia Selective agar plated medium (USP 60)
Click to view available options
:
85mm plate
Description
Burkholderia.cepacia Selective Agar Plated Medium (USP 60) is recommended for the qualitative procedures for the selective and differential isolation of B. cepacia complex in products, especially those for inhalation use of non-sterile aqueous preparations for oral, oromucosal, cutaneous or nasal use.
Confidently detect the presence of B. cepacia in nonsterile medicine with a complete workflow for USP 60 requirements which satisfies all regulations.
- Compliant formulation according to USP 60
- Quality tested to meet USP 60 specifically for Pharma customers
- Part of a complete workflow which include DCM and QCO
Specifications
Specifications
| Description | Burkholderia cepacia Selective (USP 60) Agar 10/PK |
| Format | Standard petri dish |
| Product Type | Agar Plate |
| For Use With (Application) | Pharmaceutical testing of non-sterile products used for nasal inhalation |
| Quantity | 10/Pk. |
| Regulatory Status | Lab use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction