Learn More
IKA™ I-MAG BR Stand Rod Holder
Stand rod holder for attachment to the bottom of the I-MAG industrial magnetic stirrer.
Brand:  IKA™ 0020110383
Additional Details : Weight : 0.01100kg
Product Code. 17553666
  • € 58.00 / Each
Estimated Shipment: 03-09-2024
to see stock.
Add to basket
Description
Description
  • Stand rod holder for attachment to the bottom of the I-MAG industrial magnetic stirrer.
  • In combination with support rod H 16 V and strap clamp RH 3, vessels can be securely fastened to the magnetic stirrer and secured against rotation.
  • This is especially helpful when working with viscous samples in vessels with a low dead weight.
Specifications
Specifications
I-MAG Industrial Magnetic Stirrer
0.17 kg
Aluminum Alloy
Stand Rod Holder
70 x 105 x 10 mm