missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neisseria gonorrhoeae Rabbit anti-Bacteria, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB29800.500uL
This item is not returnable.
View return policy
Description
Rabbit polyclonal antibody raised against Neisseria gonorrhoeae .
- High specificity
- High reactivity
Specifications
| Neisseria gonorrhoeae | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Neisseria gonorrhoeae . | |
| In 0.01 M Phospate buffered saline, pH 7.2 (0.09% sodium azide). | |
| Neisseria gonorrhoeae whole cells. | |
| RUO | |
| This antibody reacts with all Neisseria gonorrhoeae antigens. May react with related microbes. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Immunohistochemistry, Lateral Flow, Western Blot | |
| Unconjugated | |
| ELISA Immunofluorescence Immunohistochemistry Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 500 μL | |
| Primary | |
| Bacteria | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction