missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salmonella paratyphi A Mouse anti-Salmonella, Clone: HM175, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13652.1mg
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against Salmonella paratyphi A.
- High specificity
- High reactivity
Specifications
| Salmonella paratyphi A | |
| Monoclonal | |
| Unconjugated | |
| ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Salmonella | |
| Liquid |
| ELISA, Immunofluorescence, Lateral Flow | |
| HM175 | |
| Mouse monoclonal antibody raised against Salmonella paratyphi A. | |
| In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
| Protein A purification | |
| RUO | |
| This antibody reacts with LPS of Salmonella paratyphi. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction