Learn More
Brady™ i75 Series R6300 Ribbon for the i7500 Label Printer
Product Code. 30279163
Click to view available options
Alternate Part Number:
i75-R6300
i75-R6302
i75-R6306
i75-R6307
Unit Size:
Each
Product Code. 30279163
Brand: Brady™ 176832
Only null left
Add to basket
Add to basket
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material.
  • i75-series ribbon is self-centering with ink-side always correctly facing the label
  • One-color ribbon for printing a single color onto a label
  • R6300 Series Resin formulation offers excellent resistance to solvents, scratches and high heat when printed on the appropriate label material from Brady™
  • Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady™
  • Combine Brady label materials with their recommended ribbon series for superior legend performance
Specifications
Product Type Printer Ribbon
Color Black
For Use With (Equipment) i7500
Material Polyester
Print Technology Ink Jet
Length (English) 328.08 yd.
Width (English) 1.57 in.
Length (Metric) 300 m
Width (Metric) 40 mm
Thickness (Metric) 4.5 μm
Dimensions (L x W) 40 mm x 300 m
For Use With (Application) Barcode Labelling, Circuit Board Labelling
Quantity 1 Ea.
Alternate Part Number i75-R6306