Learn More
Brady™ i75 Series R6400 Ribbon for the i7500 Label Printer
Product Code. p-200109832
Click to view available options
Alternate Part Number:
i75-R6400
i75-R6402
i75-R6406
i75-R6407
Unit Size:
Each
Product Code. 30279169
Brand: Brady™ 176763
Only null left
Add to basket
Add to basket
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material.
  • i75-series ribbon is self-centering with ink-side always correctly facing the label
  • One-color ribbon for printing a single color onto a label
  • R6400 Series High-Density Resin Formulation offers excellent solvent, scratch and heat resistance when printed on the appropriate label material from Brady™
  • Recommended for use with materials designed to resist chemicals, such as B-492 Frozen Surface Polyester and B-481 Chemical-Resistant Polyester
  • Combine Brady label materials with their recommended ribbon series for superior legend performance
Specifications
Product Type Printer Ribbon
Color Black
For Use With (Equipment) i7500
Material Polyester
Print Technology Ink Jet
Length (English) 328.08 yd.
Width (English) 1.57 in.
Length (Metric) 300 m
Width (Metric) 40 mm
Thickness (Metric) 4.5 μm
Dimensions (L x W) 40 mm x 300 m
Quantity 1 Ea.
Alternate Part Number i75-R6406