missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IA-2/PTPRN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 433.00 - € 572.00
Specifications
| Antigen | IA-2/PTPRN |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18449220
|
Novus Biologicals
NBP1-83128-25ul |
25 μL |
€ 433.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18231236
|
Novus Biologicals
NBP1-83128 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IA-2/PTPRN Polyclonal specifically detects IA-2/PTPRN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IA-2/PTPRN | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| CA 512, EC 3.1.3.48, FLJ16131, IA2, IA-2, IA-2/PTP, ICA 512, ICA3, ICA512, islet cell antigen 2, Islet cell antigen 512, Islet cell autoantigen 3, protein tyrosine phosphatase, receptor type, N, protein tyrosine phosphatase-like N, PTP IA-2, receptor-type tyrosine-protein phosphatase-like N, R-PTP-N | |
| PTPRN | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Immune System Diseases, Immunology, Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5798 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HSYGDLPGPSPAQLFQDSGLLYLAQELPAPSRARVPRLPEQGSSSRAEDSPEGYEKEGLGDRGEKPASPAVQPDAALQRLAAVLAGYGVELRQLTPEQLSTLLTLLQLLPKGA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title