missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IA-2/PTPRN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59411
This item is not returnable.
View return policy
Description
IA-2/PTPRN Polyclonal specifically detects IA-2/PTPRN in Human samples. It is validated for Western Blot.
Specifications
| IA-2/PTPRN | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CA 512, EC 3.1.3.48, FLJ16131, IA2, IA-2, IA-2/PTP, ICA 512, ICA3, ICA512, islet cell antigen 2, Islet cell antigen 512, Islet cell autoantigen 3, protein tyrosine phosphatase, receptor type, N, protein tyrosine phosphatase-like N, PTP IA-2, receptor-type tyrosine-protein phosphatase-like N, R-PTP-N | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Equine: 92%; Bovine: 85%; Pig: 85%; Guinea pig: 84%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q7KZS4 | |
| PTPRN | |
| Synthetic peptides corresponding to PTPRN(protein tyrosine phosphatase, receptor type, N) The peptide sequence was selected from the middle region of PTPRN. Peptide sequence EVQQVPSPVSSEPPKAARPPVTPVLLEKKSPLGQSQPTVAGQPSARPAAE. | |
| 100 μL | |
| Immune System Diseases, Immunology, Lipid and Metabolism | |
| 5798 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction