missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IA-Alkyne

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
IA-Alkyne is a broad spectrum cysteine reactive probe. Covalently binds cysteine residues in proteins. Can be utilized for covalent fragment-based ligand discovery (FBLD) and to identify Zn2+-binding cysteines. Used as a click-chemistry handle to attach a fluorophore or biotin to reactive cysteines in proteins for visualization by in-gel fluorescence or identification by mass spectrometry, respectively. Covalently binds cysteine residues in and activates TRPA1 channels.
Specifications
Specifications
| Chemical Name or Material | N-5-Hexyn-1-yl-2-iodoacetamide |
| CAS | 930800-38-7 |
| Quantity | 5 mg |
| Target | General Laboratory Research Reagent |
| Molecular Formula | C8H12INO |
| Formula Weight | 1919 |
| Purity | 97% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction