missing translation for 'onlineSavingsMsg'
Learn More

IACS-9571 hydrochloride, MedChemExpress

Product Code. 16430969
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16430969

Brand: Medchem Express HY102000B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IACS-9571 (ASIS-P040) hydrochloride is a potent and selective inhibitor of TRIM24 and BRPF1, with an IC50 of 8 nM for TRIM24, and Kds of 31 nM and 14 nM for TRIM24 and BRPF1, respectively.

IACS-9571 (ASIS-P040) hydrochloride is a potent and selective inhibitor of TRIM24 and BRPF1, with an IC50 of 8 nM for TRIM24, and Kds of 31 nM and 14 nM for TRIM24 and BRPF1, respectively.

Specifications

Chemical Name or Material IACS-9571 hydrochloride
CAS 2319611-93-1
Color Brown
Purity Grade Notes Research
Molecular Formula C32H43ClN4O8S
Quantity 1 mg
Synonym ASIS-P040 hydrochloride
Solubility Information DMSO : 125 mg/mL (184.03 mM; Need ultrasonic and warming) ∣H2O : 10 mg/mL (14.72 mM; Need ultrasonic)
SMILES [H]Cl.O=S(C1=CC=C(OC)C(OC)=C1)(NC2=C(OC3=CC(OCCC)=CC(OCCCCN(C)C)=C3)C=C(N4C)C(N(C)C4=O)=C2)=O
Molecular Weight (g/mol) 679.22
Formula Weight 679.22
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.