missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IARS2 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18443R
This item is not returnable.
View return policy
Description
IARS2 Polyclonal antibody specifically detects IARS2 in Human samples. It is validated for Western Blot
Specifications
| IARS2 | |
| Polyclonal | |
| Western Blot | |
| EC 6.1.1, EC 6.1.1.5, FLJ10326, ILERS, isoleucine tRNA ligase 2, mitochondrial, Isoleucine--tRNA ligase, isoleucine-tRNA synthetase 2, mitochondrial, isoleucyl-tRNA synthetase 2, mitochondrial, isoleucyl-tRNA synthetase, mitochondrial, mitochondrial isoleucine tRNA synthetase | |
| Recognizes region between residues Between Between 962 and 1012.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55699 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction