Learn More
IKA™ IB R RO 15 eco
Plastic bath designed for magnetic stirrer RO 15 size L.
Brand: IKA™ 0020006341
Additional Details : Weight : 6.67500kg
Description
- Max. temperature: 80°C; water only
- Inner dimensions: 674 x 317 x 98 mm
- Outer dimensions: 722 x 365 x 165 mm
Specifications
25 L | |
80°C | |
6.675 kg | |
Plastic |
Magnetic stirrer | |
Plastic Bath | |
Outer: 722 x 365 x 165 mm; Inner: 674 x 317 x 98 mm |