missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBRDC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00255488-D01P
This item is not returnable.
View return policy
Description
IBRDC2 Polyclonal antibody specifically detects IBRDC2 in Human samples. It is validated for Western Blot
Specifications
| IBRDC2 | |
| Polyclonal | |
| Western Blot 1:500 | |
| AAH63311.1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 255488 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| bA528A10.3, E3 ubiquitin-protein ligase RNF144B, EC 6.3.2, EC 6.3.2.-, IBR domain containing 2, IBR domain-containing protein 2, IBRDC2, KIAA0161, p53-inducible RING finger protein, P53RFP, PIR2, ring finger protein 144BMGC71786 | |
| RNF144B (AAH63311.1, 1 a.a. - 302 a.a.) full-length human protein. MGSAGRLHYLAMTAENPTPGDLAPAPLITCKLCLCEQSLDKMTTLQECQCIFCTACLKQYMQLAIREGCGSPITCPDMVCLNHGTLQEAEIACLVPVDQFQLYQRLKFEREVHLDPYRTWCPVADCQTVCPVASSDPGQPVLVECPSCHLKFCSCCKDAWHAEVSCRDSQPIVLPTEHRALFGTDAEAPIKQCPVCRVYIERNEGCAQMMCKNCKHTFCWYCLQNLDNDIFLRHYDKGPCRNKLGHSRASVMWNRTQVVGILVGLGIIALVTSPLLLASPCIICCVCKSCRGKKKKHDPSTT | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction