missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBRDC2 Antibody (OTI9H10), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72470MFV610
This item is not returnable.
View return policy
Description
IBRDC2 Monoclonal antibody specifically detects IBRDC2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| IBRDC2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 255488 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| OTI9H10 | |
| Flow Cytometry | |
| bA528A10.3, E3 ubiquitin-protein ligase RNF144B, EC 6.3.2, EC 6.3.2.-, IBR domain containing 2, IBR domain-containing protein 2, IBRDC2, KIAA0161, p53-inducible RING finger protein, P53RFP, PIR2, ring finger protein 144BMGC71786 | |
| Human recombinant protein fragment corresponding to amino acids 1-256 of human RNF144B (NP_877434) produced in E. coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction