Learn More
Ibrutinib-biotin, MedChemExpress
Product Code. 16445697
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16445697 5 mg 5mg
16422958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16445697 Supplier Medchem Express Supplier No. HY100342
MedChemExpress Ibrutinib-biotin is a probe that consists of Ibrutinib linked to biotin via a long chain linker, extracted from patent WO2014059368A1 Compound 1-5, has an IC50 of 0.755-1.02 nM for BTK.
Ibrutinib-biotin is a probe that consists of Ibrutinib linked to biotin via a long chain linker, extracted from patent WO2014059368A1 Compound 1-5, has an IC50 of 0.755-1.02 nM for BTK.
Specifications
Chemical Name or Material Ibrutinib-biotin
CAS 1599432-18-4
Color Colorless
Purity Grade Notes Research
Molecular Formula C56H80N12O9S
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (45.56 mM; Need ultrasonic)
SMILES O=C(CCCC(NCCCOCCOCCOCCCNC(CCCC[C@@H]1SC[C@H](N2)[C@@H]1NC2=O)=O)=O)NCCCCN(C)C/C=C/C(N3CCC[C@@H](N4N=C(C5=CC=C(OC6=CC=CC=C6)C=C5)C7=C(N)N=CN=C74)C3)=O
Molecular Weight (g/mol) 1097.37
Formula Weight 1097.37
Percent Purity 98.04%
Grade Research
Physical Form Oil
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only