missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IBSP Polyclonal Antibody
GREENER_CHOICE

Product Code. 17207683 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17207683 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17207683 Supplier Invitrogen™ Supplier No. PA5114915

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Bone Sialoprotein.

The protein encoded by this gene is a major structural protein of the bone matrix. It constitutes approximately 12% of the noncollagenous proteins in human bone and is synthesized by skeletal-associated cell types, including hypertrophic chondrocytes, osteoblasts, osteocytes, and osteoclasts. The only extraskeletal site of its synthesis is the trophoblast. This protein binds to calcium and hydroxyapatite via its acidic amino acid clusters, and mediates cell attachment through an RGD sequence that recognizes the vitronectin receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IBSP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ibsp
Gene Accession No. P13839, P21815, Q61711
Gene Alias BNSP; Bone sialoprotein 2; bone sialoprotein II; Bsp; BSP II; Bsp2; BSPII; BSP-II; Cell-binding sialoprotein; IBSP; integrin binding sialoprotein; integrin-binding sialoprotein; SP-II
Gene Symbols Ibsp
Host Species Rabbit
Immunogen A synthesized peptide derived from human IBSP(Accession P21815), corresponding to amino acid residues E166-E216.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15891, 24477, 3381
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.