missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBTK Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50033H
This item is not returnable.
View return policy
Description
IBTK Polyclonal antibody specifically detects IBTK in Human samples. It is validated for Western Blot
Specifications
| IBTK | |
| Polyclonal | |
| Western Blot | |
| Bruton agammaglobulinemia tyrosine kinase inhibitor, BTK-binding protein, BTKI, DKFZP564B116, IBtk, inhibitor of Bruton agammaglobulinemia tyrosine kinase, inhibitor of Bruton tyrosine kinase, inhibitor of Bruton's tyrosine kinase, inhibitor of Bruton's tyrosine kinase-alpha, inhibitor of Bruton's tyrosine kinase-beta, inhibitor of Bruton's tyrosine kinase-gamma, KIAA1417, MGC142256, MGC142258, RP1-93K22.1 | |
| The immunogen for this product maps to a region between residue 1050 and 1100 of human Inhibitor of Bruton Agammaglobulinemia Tyrosine Kinase, using the numbering given in entry NP_056340.2 (GeneID 25998). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 25998 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction