missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1/CD54 Antibody (141032), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne FAB5831L
This item is not returnable.
View return policy
Description
ICAM-1/CD54 Monoclonal antibody specifically detects ICAM-1/CD54 in Rat samples. It is validated for Western Blot, Flow Cytometry
Specifications
| ICAM-1/CD54 | |
| Monoclonal | |
| DyLight 550 | |
| BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
| Mouse myeloma cell line NS0-derived recombinant rat ICAM-1/CD54, Gln28-Thr493 (predicted), Accession # Q00238 | |
| 0.5 mL | |
| Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers | |
| 3383 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry | |
| 141032 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction