missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1/CD54 Antibody (3E2B) - Azide and BSA Free, Novus Biologicals™
Hamster Monoclonal Antibody has been used in 1 publication
€ 186.00 - € 519.00
Specifications
| Antigen | ICAM-1/CD54 |
|---|---|
| Clone | 3E2B |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:10 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:200, Inhibition Activity, CyTOF-ready, Neutralization |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Inhibition Assays, CyTOF, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18420452
|
Novus Biologicals
NBP2-22540-100ug |
100 ug |
€ 186.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18751744
|
Novus Biologicals
NBP2-22540 |
€ 519.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Inhibition Activity, CyTOF-ready, Block/Neutralize.Specifications
| ICAM-1/CD54 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Inhibition Assays, CyTOF, Neutralization | |
| Unconjugated | |
| Hamster | |
| Human, Mouse | |
| 3383 | |
| Mouse ICAM-1 (CD54) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 3E2B | |
| Western Blot 1:10 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:200, Inhibition Activity, CyTOF-ready, Neutralization | |
| Monoclonal | |
| Purified | |
| RUO | |
| BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
| ICAM1 | |
| IgG | |
| Protein G purified | |
| This antibody reacts with Mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title