missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1/CD54 Antibody (bersanlimab), FITC, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27998F
This item is not returnable.
View return policy
Description
ICAM-1/CD54 Monoclonal antibody specifically detects ICAM-1/CD54 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ICAM-1/CD54 | |
| Monoclonal | |
| FITC | |
| BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
| ICAM1/ CD54 | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers | |
| 3383 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| bersanlimab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction