missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1/CD54 Antibody (ICAM1/9379R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 291.00 - € 519.00
Specifications
| Antigen | ICAM-1/CD54 |
|---|---|
| Clone | ICAM1/9379R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671004
|
Novus Biologicals
NBP3-23796-100ug |
100 μg |
€ 519.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650535
|
Novus Biologicals
NBP3-23796-20ug |
20 μg |
€ 291.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICAM-1/CD54 Monoclonal antibody specifically detects ICAM-1/CD54 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ICAM-1/CD54 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 3383 | |
| IgG κ | |
| Protein A or G purified |
| ICAM1/9379R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
| IL-1B activated human vascular endothelial cells expressing ICAM-1/CD54. | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title